Learn More
Bioanalytical Systems Inc EC VOLTAMMETRY CELL
SDP
Catalog No. 50854512
Encompass_Preferred
Cell, Voltammetry; BASi VC-2; With PTFE Electrode Holder, PTFE Tubing, and Platinum Wire Auxiliary Electrode *Does not ship to Puerto Rico
Catalog No. 50-854-512 Supplier Bioanalytical Systems Inc Supplier No. MF1052
Cell, Voltammetry; BASi VC-2; With PTFE Electrode Holder, PTFE Tubing, and Platinum Wire Auxiliary Electrode *Does not ship to Puerto Rico