Learn More
Macherey-Nagel MACHEREY-NAGEL EC 150/2 NUCLEODUR HILIC HPLC Column, 5 µm Particle Size, 150 mm Length, 2.0 mm ID
Catalog No. NC2871665
Encompass_Preferred
The MACHEREY-NAGEL EC 150/2 NUCLEODUR HILIC HPLC column is engineered for high-performance separation of polar analytes through Hydrophilic Interaction Chromatography (HILIC). Its unique fully porous silica stationary phase and 5 µm particle size ensure reproducible, stable performance, setting it apart from conventional methods. Ideal for scenarios where reversed-phase techniques fall short, it optimizes analytical capabilities for researchers seeking precise results in complex sample matrices.
Catalog No. NC2871665 Supplier Macherey-Nagel Supplier No. 760553.20
The MACHEREY-NAGEL EC 150/2 NUCLEODUR HILIC HPLC column is engineered for high-performance separation of polar analytes through Hydrophilic Interaction Chromatography (HILIC). Its unique fully porous silica stationary phase and 5 µm particle size ensure reproducible, stable performance, setting it apart from conventional methods. Ideal for scenarios where reversed-phase techniques fall short, it optimizes analytical capabilities for researchers seeking precise results in complex sample matrices.