Learn More
Medchemexpress LLC HY-12309 5mg Medchemexpress, EC23 CAS:104561-41-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123095MG
Encompass_Preferred
Medchemexpress, HY-12309 5mg EC23 CAS:104561-41-3 EC23 (AGN 190205) is a stable synthetic retinoid analogue and induces neuronal differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9645