Learn More
STEMCELL Technologies EC23, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 73102
Encompass
EC23 is a retinoic acid receptor (RAR) agonist with pan-RAR activity (EC₅₀: RARα 41 nM, RARβ 0.5 nM, RARγ 0.4 nM), while having no appreciable activity for retinoid X receptors (RXR; EC₅₀> 10μM for all; Gambone et al.). It is a photostable synthetic analog of all-trans retinoic acid (ATRA; Christie et al. 2008). EC23 also weakly activates aryl hydrocarbon receptor (Gambone et al.).DIFFERENTIATION· Induces neural differentiation of human pluripotent stem cells, similarly to ATRA (Christie et al. 2010; Clemens et al.).· Induces neuronal differentiation of human fetal neural progenitor cell line ReNcell 197VM (Christie et al. 2010).
Catalog No. 50-197-6026