Learn More
Medchemexpress LLC Ec330 | 2016795-77-8 | 99.9% | 462.57 g/mol | C30H32F2O2 | 25 MG
SDP
Catalog No. 5000212612
Encompass_Preferred
EC330 is a small-molecule leukemia inhibitory factor (LIF) inhibitor supplied as a solid powder for research use. The compound has CAS 2016795-77-8, molecular formula C30H32F2O2, molecular weight 462.57 g/mol, and is provided with high purity suitable for biochemical and cell-based studies. For research use only; not for human or diagnostic applications.
  • High purity suitable for biochemical and cell-based assays.
  • Solid powder form for easy handling and storage.
  • Known molecular formula and molecular weight for accurate dosing.
  • Applicable to in vitro and in vivo research studies.
  • Includes batch-specific certificate of analysis for quality verification.
Catalog No. 50-002-12612 Supplier Medchemexpress LLC Supplier No. HY10094925MG
EC330 is a small-molecule leukemia inhibitory factor (LIF) inhibitor supplied as a solid powder for research use. The compound has CAS 2016795-77-8, molecular formula C30H32F2O2, molecular weight 462.57 g/mol, and is provided with high purity suitable for biochemical and cell-based studies. For research use only; not for human or diagnostic applications.
  • High purity suitable for biochemical and cell-based assays.
  • Solid powder form for easy handling and storage.
  • Known molecular formula and molecular weight for accurate dosing.
  • Applicable to in vitro and in vivo research studies.
  • Includes batch-specific certificate of analysis for quality verification.