Learn More
Medchemexpress LLC HY-120142 5mg Medchemexpress, EC359 CAS:2012591-09-0 Purity:>98%
SDP
Catalog No. 501969584
Encompass_Preferred
Medchemexpress, HY-120142 5mg EC359 CAS:2012591-09-0 EC359 is a potent, selective, high affinity and orally active leukemia inhibitory factor receptor (LIFR) inhibitor with a Kd of 10.2 nM, which directly interacts with LIFR to effectively block LIF/LIFR interactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9584 Supplier Medchemexpress LLC Supplier No. HY1201425MG
Medchemexpress, HY-120142 5mg EC359 CAS:2012591-09-0 EC359 is a potent, selective, high affinity and orally active leukemia inhibitory factor receptor (LIFR) inhibitor with a Kd of 10.2 nM, which directly interacts with LIFR to effectively block LIF/LIFR interactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.