Learn More
Medchemexpress LLC HY-135653 10mg , EC5026 CAS:1809885-32-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13565310MG
Encompass_Preferred
HY-135653 10mg EC5026 CAS: 1809885-32-2 EC5026 (BPN-19186) is a first-in-class, non-opioid and orally active soluble Epoxide Hydrolase (sEH) inhibitor. EC5026 shows efficacy for inflammatory and neuropathic pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0773