Learn More
Medchemexpress LLC HY-135653 5mg Medchemexpress, EC5026 CAS:1809885-32-2 Purity:>98%
SDP
Catalog No. 501969882
Encompass_Preferred
Medchemexpress, HY-135653 5mg EC5026 CAS:1809885-32-2 EC5026 (BPN-19186) is a first-in-class, potent and orally active soluble Epoxide Hydrolase (sEH) inhibitor. Inhibition of sEH treats pain by stabilizing natural analgesic and anti-inflammatory mediators. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9882 Supplier Medchemexpress LLC Supplier No. HY1356535MG
Medchemexpress, HY-135653 5mg EC5026 CAS:1809885-32-2 EC5026 (BPN-19186) is a first-in-class, potent and orally active soluble Epoxide Hydrolase (sEH) inhibitor. Inhibition of sEH treats pain by stabilizing natural analgesic and anti-inflammatory mediators. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.