Learn More
TARGETMOL CHEMICALS INC ECABAPIDE 5MG
SDP
Catalog No. 502576986
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg and bulk. Please contact Fisher for quotes. Ecabapide (DQ-2511) is a novel gastrointestinal motility promoter that significantly enhances gastric emptying in SHRSP.Ecabapide activates cyclic GMP-dependent housekeeping Cl- channels in rabbit gastric wall cells. purity: 99%
Catalog No. 50-257-6986 Supplier TARGETMOL CHEMICALS INC Supplier No. T315975MG
Also available in 1mg 10mg 25mg 50mg and bulk. Please contact Fisher for quotes. Ecabapide (DQ-2511) is a novel gastrointestinal motility promoter that significantly enhances gastric emptying in SHRSP.Ecabapide activates cyclic GMP-dependent housekeeping Cl- channels in rabbit gastric wall cells. purity: 99%