Learn More
Selleck Chemical LLC Ecabet sodium S4853-25mg
SDP
Catalog No. 503110697
Encompass_Preferred
Ecabet sodium (TA-2711 TA-2711E) is a widely employed mucoprotective agent for the treatment of gastric ulcers Ecabet sodium inhibits the ability of Helicobacter pylori to induce neutrophil production of reactive oxygen species and interleukin-8 Ecabet sodium can also reduce apoptosis
Catalog No. 50-311-0697 Supplier Selleck Chemical LLC Supplier No. S485325MG
Add to Cart
Add to Cart
Ecabet sodium (TA-2711 TA-2711E) is a widely employed mucoprotective agent for the treatment of gastric ulcers Ecabet sodium inhibits the ability of Helicobacter pylori to induce neutrophil production of reactive oxygen species and interleukin-8 Ecabet sodium can also reduce apoptosis