Learn More
Medchemexpress LLC Ecallantide (DX-88) trifluoroacetate | 00-00-0 | 7053.81 g/mol (free base) | C305H442N88O91S8 · xC2HF3O2 | 5 MG
SDP
Catalog No. 5000264079
Encompass_Preferred
Ecallantide TFA is a recombinant plasma kallikrein inhibitor supplied as the trifluoroacetate salt for research use. It inhibits kallikrein-mediated formation of bradykinin and is used in laboratory studies of angioedema and bradykinin-related pathways. Provided in defined small-scale packages for analytical and preclinical research; not for human use.
  • Specific recombinant plasma kallikrein inhibitor.
  • Trifluoroacetate salt formulation for stability and handling.
  • Available in small, defined package sizes suitable for research.
  • Characterized molecular weight and molecular formula provided.
  • Intended for research use only; not for clinical or human use.
  • Applicable to studies of bradykinin-mediated signaling and angioedema models.
Catalog No. 50-002-64079 Supplier Medchemexpress LLC Supplier No. HY108814A5MG
Ecallantide TFA is a recombinant plasma kallikrein inhibitor supplied as the trifluoroacetate salt for research use. It inhibits kallikrein-mediated formation of bradykinin and is used in laboratory studies of angioedema and bradykinin-related pathways. Provided in defined small-scale packages for analytical and preclinical research; not for human use.
  • Specific recombinant plasma kallikrein inhibitor.
  • Trifluoroacetate salt formulation for stability and handling.
  • Available in small, defined package sizes suitable for research.
  • Characterized molecular weight and molecular formula provided.
  • Intended for research use only; not for clinical or human use.
  • Applicable to studies of bradykinin-mediated signaling and angioedema models.