Learn More
Medchemexpress LLC HY-16182A 500mg , Ecamsule (disodium) CAS:90458-75-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16182A500MG
Encompass_Preferred
HY-16182A 500mg Ecamsule (disodium) CAS: 90458-75-6 Ecamsule disodium is a broad-spectrum UVA filter that can be used in sunscreen product. Ecamsule reduces biological damage caused by solar radiation such as pyrimidine dimer formation, p53 protein accumula-tion, or collagenase 2 expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1493