Learn More
Grainger Eccentric Reducer Coupling SS 4 x 2 in
Catalog No. 502749719
Encompass_Preferred
The Blucher Eccentric Reducer Coupling is a stainless steel pipe fitting that connects pipes of different diameters. It has an eccentric design that offsets the centerlines of the larger and smaller pipe connections to provide a smooth transition. This 2" x 1-1/2" reducer coupling has 150# flanged ends and is made of 304 stainless steel for corrosion resistance. It can handle temperatures up to 850°F and pressures up to 285 psi. The eccentric design prevents turbulence and minimizes pressure loss. This high-quality reducer coupling provides a reliable transition between different pipe sizes in industrial piping systems.
Catalog No. 50-274-9719 Supplier Grainger Supplier No. 14P006
The Blucher Eccentric Reducer Coupling is a stainless steel pipe fitting that connects pipes of different diameters. It has an eccentric design that offsets the centerlines of the larger and smaller pipe connections to provide a smooth transition. This 2" x 1-1/2" reducer coupling has 150# flanged ends and is made of 304 stainless steel for corrosion resistance. It can handle temperatures up to 850°F and pressures up to 285 psi. The eccentric design prevents turbulence and minimizes pressure loss. This high-quality reducer coupling provides a reliable transition between different pipe sizes in industrial piping systems.