Learn More
Grainger Eccentric Reducer Coupling SS 4 x 3 in
Catalog No. 502748701
Encompass_Preferred
The Anvil Eccentric Reducer Coupling is a ductile iron pipe fitting that reduces pipe size while allowing adjustment of the outlet position. It has a 4 inch inlet and a 3 inch outlet, and allows up to 5 degrees of outlet rotation for proper alignment. The coupling is rated for pressures up to 350 psi and temperatures up to 250°F. It is made in the USA and conforms to ASTM A536 standards. The coupling has a fusion-bonded epoxy coating for corrosion resistance.
Catalog No. 50-274-8701 Supplier Grainger Supplier No. 14P007
The Anvil Eccentric Reducer Coupling is a ductile iron pipe fitting that reduces pipe size while allowing adjustment of the outlet position. It has a 4 inch inlet and a 3 inch outlet, and allows up to 5 degrees of outlet rotation for proper alignment. The coupling is rated for pressures up to 350 psi and temperatures up to 250°F. It is made in the USA and conforms to ASTM A536 standards. The coupling has a fusion-bonded epoxy coating for corrosion resistance.