Learn More
MOLTOX Molecular Toxicology Inc ECDisc WP2 trp, Consists of lyophilized & stabilized E.Coli strain WP2 trp cells, For use as culture inoculum, 10 discs/vial, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 72003L
Encompass
Escherichia coli strains (WP2 derivatives) for use in the bacterial mutagenesis assay described by Green & Muriel, Mutat. Res. 38: 3, 1976. Most commonly employed bacterial strains for use in mutagenicity testing are provided in convenient lyophilized disc format. Strains are periodically characterized and verified for diagnostic performance. Confirmed strains are lyophilized in modified ATCC Reagent 18. After preservation, samples are cultured and their phenotypes and responses to diagnostic mutagens are determined. Strains are accompanied by Quality Control statements that include the results of phenotype confirmation tests, diagnostic mutagen response and viability assessment. Culture instructions are included with the QC Statement. The bacterial strains contained in STDiscs™ and ECDiscs™ are potential etiologic agents and are intended for use only by those skilled in the safe handling of potentially infectious agents.
Catalog No. 50-948-945