Learn More
AdipoGen Ecdysterone
SDP
Supplier:  AdipoGen CDXE0225M005
Encompass_Preferred
Chemical. CAS 5289-74-7. Formula C27H44O7. MW 480.63. Synthetic. A member of the ecdysteroid family. Ecdysone receptor EcR agonist. More potent than ecdysone. Induces the expression of genes coding for proteins that the larva requires, and it causes chromosome puffs sites of high expression to form in polytene chromosomes. Plays a key role in insect development, cell proliferaton, growth and apoptosis by controlling gene expression involved in moulting and metamorphosis. It acts through a heterodimeric receptor comprising the ecdysone receptor and the ultraspiracle proteins USP. Used for controlled gene expression in scientific research, agriculture and medicine. Used for the development of selective insect growth regulators for use as environmentally benign insecticides. Shows biological effects on mammalian species, including antiepileptic, antidiabetic, antiobesity, ROS-inhibiting activity.ht produces vitamin D3. UV/Vis lambdamax 271, 282, 293nm.
Catalog No. 50-256-0966