Learn More
Apexbio Technology LLC Ecdysterone 5289-74-7 20mg
SDP
Supplier:  Apexbio Technology LLC N135020
Encompass_Preferred
Ecdysterone (CAS 5289-74-7) is a naturally occurring phytoecdysteroid predominantly isolated from Cyanotis arachnoidea It functions as an agonist of the ecdysone receptor modulating gene expression associated with growth protein synthesis and cellular differentiation Ecdysterone has been investigated for its anabolic effects without significant androgenic activity making it a subject of interest in muscle biology and metabolic research Soluble in methanol and DMSO this compound is suitable for a range of in vitro and in vivo studies examining molecular mechanisms underlying cellular growth and adaptation Store Ecdysterone in a cool dry and sealed environment
Catalog No. 50-000-90697