Learn More
Selleck Chemical LLC eCF506 S0791-5MG
SDP
Catalog No. 5000042596
Encompass_Preferred
eCF506 is a potent selective and orally bioavailable inhibitor of Src tyrosine kinase with IC50 0 5 nM
Catalog No. 50-000-42596 Supplier Selleck Chemical LLC Supplier No. S07915MG
eCF506 is a potent selective and orally bioavailable inhibitor of Src tyrosine kinase with IC50 0 5 nM