Learn More
World Precision Instrument ECG Analysis Module
SDP
Catalog No. 502539212
Encompass
  • License for ECG Analysis module in LabTrax-4
  • Measure R-R, PR, QT, QR and QTc intervals, QRS, T, P and TP durations, P, Q, R, S, T amplitudes and ST elevation, etc...
  • Specific analysis templates are included to accurately delineate PQRST onset, durations and amplitudes
  • Custom templates can also be created based on unique ECG profiles and saved to a library for future use
  • Beat averaging and classification
  • Outlier removal based on abnormal R-R intervals, heart rate, noise and activity
  • Easily extract source data and average data as images or text files
  • Plug and play compatible with all iWorx data acquisition
  • Optional ASCII Text Import Module is also available that enables import of ECG data in text or edf format that was recorded by any other acquisition program
  • The software unlock key will based on the user’s name
Catalog No. 50-253-9212 Supplier World Precision Instrument Supplier No. 505198
  • License for ECG Analysis module in LabTrax-4
  • Measure R-R, PR, QT, QR and QTc intervals, QRS, T, P and TP durations, P, Q, R, S, T amplitudes and ST elevation, etc...
  • Specific analysis templates are included to accurately delineate PQRST onset, durations and amplitudes
  • Custom templates can also be created based on unique ECG profiles and saved to a library for future use
  • Beat averaging and classification
  • Outlier removal based on abnormal R-R intervals, heart rate, noise and activity
  • Easily extract source data and average data as images or text files
  • Plug and play compatible with all iWorx data acquisition
  • Optional ASCII Text Import Module is also available that enables import of ECG data in text or edf format that was recorded by any other acquisition program
  • The software unlock key will based on the user’s name