Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterprise Technology Solutions ECG Snap Electrode 50 mm x 55
SDP

Catalog No. 502886037
Encompass_Preferred

ECG Snap Electrode 50 mm x 55 mm - Adult 12/50/cs

Catalog No. 50-288-6037 Supplier Enterprise Technology Solutions Supplier No. ETS7103
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ECG Snap Electrode 50 mm x 55 mm - Adult 12/50/cs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.