Learn More
MilliporeSigma™ ech2o™ Lamp kit
Catalog No. IQ7XUVKT1 Shop All MilliporeSigma Products
Change view
Click to view available options
Lamp kit
Catalog No.
IQ7XUVKT1 Lamp kit
1 options
Catalog No. IQ7XUVKT1 Supplier MilliporeSigma™ Supplier No. IQ7XUVKT1
Add to Cart
Add to Cart
MilliporeSigma™ ech2o™ Lamp Kit for Milli-Q™ IQ 7003/05/10/15 Water Purification Systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Milli-Q™ IQ 7003/7005/7010/7015 Systems
Includes Lamp kit
Electrical Requirements Voltage: 100 to 240 V ±10%; Frequency 50 to 60 Hz ±10%
Declaration of CE and cUL Conformity: The Milli-Q™ IQ 7003, IQ 7005, IQ 7010 and IQ 7015 systems have been tested by an independent and accredited company for compliance with CE directives related to safety and electromagnetic compatibility. The report can be consulted on demand. In addition, the system is built using components and practices recommended by UL and has been cUL marked. The registration can be verified on the UL web site