Learn More
Aviva Systems Biology Corp ECHDC2 Antibody - middle region (ARP48828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ECHDC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ECHDC2. Alternate names: FLJ10948ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59930