Learn More
Aviva Systems Biology Corp ECHDC3 Antibody - N-terminal region (ARP48911_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48911_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ECHDC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ECHDC3. Alternate names: RP11-401F24.3, FLJ20909ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59958