Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Analytical Sales & Services Echelon C18, 1.7μm, 10cm x 2.1mm

Catalog No. 5000805010
Encompass

Echelon C18, 1.7μm, 10cm x 2.1mm 100Å,

Catalog No. 50-008-05010 Supplier Analytical Sales & Services Supplier No. ADV8117
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Echelon C18, 1.7μm, 10cm x 2.1mm 100Å,

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.