Learn More
Analytical Sales & Services Echelon C18 Analytical Column. 4µm, 10cm x 3.0mm ID
Catalog No. 5000805580
Encompass
Echelon C18 Analytical Column. 4µm, 10cm x 3.0mm ID
Catalog No. 50-008-05580 Supplier Analytical Sales & Services Supplier No. ADV8005
Echelon C18 Analytical Column. 4µm, 10cm x 3.0mm ID