Learn More
Analytical Sales & Services Echelon C18 Analytical Column. 4µm, 25cm x 4.6mm ID
Catalog No. 5000805636
Encompass
Echelon C18 Analytical Column. 4µm, 25cm x 4.6mm ID
Catalog No. 50-008-05636 Supplier Analytical Sales & Services Supplier No. ADV8003
Add to Cart
Add to Cart
Echelon C18 Analytical Column. 4µm, 25cm x 4.6mm ID