Learn More
Analytical Sales & Services Echelon C18 Analytical Column. 4µm, 5cm x 4.6mm ID
Catalog No. 5000806642
Encompass
Echelon C18 Analytical Column. 4µm, 5cm x 4.6mm ID
Catalog No. 50-008-06642 Supplier Analytical Sales & Services Supplier No. ADV8000
Add to Cart
Add to Cart
Echelon C18 Analytical Column. 4µm, 5cm x 4.6mm ID