Learn More
Analytical Sales & Services Echelon C18 Microbore Column. 3µm, 5cm x 0.3mm ID
Catalog No. 5000805657
Encompass
Echelon C18 Microbore Column. 3µm, 5cm x 0.3mm ID
Catalog No. 50-008-05657 Supplier Analytical Sales & Services Supplier No. ADV0503183
Echelon C18 Microbore Column. 3µm, 5cm x 0.3mm ID