Learn More
Analytical Sales & Services Echelon C18 Microbore Column. 4µm, 10cm x 0.5mm ID
Catalog No. 5000805899
Encompass
Echelon C18 Microbore Column. 4µm, 10cm x 0.5mm ID
Catalog No. 50-008-05899 Supplier Analytical Sales & Services Supplier No. ADV1005185
Echelon C18 Microbore Column. 4µm, 10cm x 0.5mm ID