Learn More
Analytical Sales & Services Echelon C18 Microbore Column. 4µm, 25cm x 0.3mm ID
Catalog No. 5000805466
Encompass
Echelon C18 Microbore Column. 4µm, 25cm x 0.3mm ID
Catalog No. 50-008-05466 Supplier Analytical Sales & Services Supplier No. ADV2503185
Echelon C18 Microbore Column. 4µm, 25cm x 0.3mm ID