Learn More
Analytical Sales & Services Echelon C18 Microbore Column. 4µm, 25cm x 1.0mm ID
Catalog No. 5000805962
Encompass
Echelon C18 Microbore Column. 4µm, 25cm x 1.0mm ID
Catalog No. 50-008-05962 Supplier Analytical Sales & Services Supplier No. ADV2510185
Echelon C18 Microbore Column. 4µm, 25cm x 1.0mm ID