Learn More
Analytical Sales & Services Echelon C18 Microbore Column. 4µm, 3cm x 0.3mm ID
Catalog No. 5000805840
Encompass
Echelon C18 Microbore Column. 4µm, 3cm x 0.3mm ID
Catalog No. 50-008-05840 Supplier Analytical Sales & Services Supplier No. ADV0303185
Add to Cart
Add to Cart
Echelon C18 Microbore Column. 4µm, 3cm x 0.3mm ID