Learn More
Analytical Sales & Services Echelon C18 Prep Column. 4µm, 10cm x 20mm ID
Catalog No. 5000805396
Encompass
Echelon C18 Prep Column. 4µm, 10cm x 20mm ID
Catalog No. 50-008-05396 Supplier Analytical Sales & Services Supplier No. ADV821020
Echelon C18 Prep Column. 4µm, 10cm x 20mm ID