Learn More
Analytical Sales & Services Echelon C18 Semi-Prep Column. 4µm, 15cm x 10mm ID
Catalog No. 5000805630
Encompass
Echelon C18 Semi-Prep Column. 4µm, 15cm x 10mm ID
Catalog No. 50-008-05630 Supplier Analytical Sales & Services Supplier No. ADV821510
Echelon C18 Semi-Prep Column. 4µm, 15cm x 10mm ID