Learn More
Cayman Chemical Echinacoside, Purity: ≥95%, CAS: 82854-37-3, 50 mg
SDP
Catalog No. 502992185
Encompass_Preferred
A phenylethanoid glycoside; scavenges DPPH radicals (EC50 = 6.6 µM in a cell-free assay); inhibits hydrogen peroxide-induced production of ROS in a cell-free assay at 10 µg/ml; reduces increases in plasma ALT, TNF-α, IL-1β, IL-16, and IL-10 levels and hepatocyte apoptosis induced by LPS/D-galactosamine in a mouse model of acute liver injury at 60 mg/kg; prevents loss of dopaminergic neurons in the substantia nigra pars compacta and decreases in striatal dopamine and DOPAC levels in a mouse model of MPTP-induced Parkinson's disease; improves motor performance in the rotarod test in the same model
Catalog No. 50-299-2185 Supplier Cayman Chemical Supplier No. 2992750 MG
A phenylethanoid glycoside; scavenges DPPH radicals (EC50 = 6.6 µM in a cell-free assay); inhibits hydrogen peroxide-induced production of ROS in a cell-free assay at 10 µg/ml; reduces increases in plasma ALT, TNF-α, IL-1β, IL-16, and IL-10 levels and hepatocyte apoptosis induced by LPS/D-galactosamine in a mouse model of acute liver injury at 60 mg/kg; prevents loss of dopaminergic neurons in the substantia nigra pars compacta and decreases in striatal dopamine and DOPAC levels in a mouse model of MPTP-induced Parkinson's disease; improves motor performance in the rotarod test in the same model