Learn More
Alpco Ltd Echinococcus Granulosus IgG ELISA
SDP
Supplier:  Alpco Ltd 20EGGHUE01
Encompass_Preferred
The Echinococcus IgG Enzyme Immunoassay Kit provides materials for the qualitative and semiquantitative determination of IgG-class antibodies to Echinococcus in human serum and plasma. FDA Registered. For In Vitro Diagnostic Use.
Catalog No. 50-305-090