Learn More
Drg International Inc Echinococcus Igg Elisa
Supplier:  Drg International Inc EIA3472
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Echinococcus in serum and plasma. Sources of infection are final hosts (i.e. dogs for E. granulosus and mainly foxes for E. multilocularis) and food contaminated with parasite eggs. After ingestion of a suitable intermediate host, the egg hatches in the small bowel and releases an oncosphere that penetrates the intestinal wall and through the circulatory system into various organs where it develops into a cyst. Echinococcus infections remain silent for years before the enlarging cysts cause symptoms in the affected organs. The DRG Echinococcus IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Echinococcus antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7568