Learn More
TARGETMOL CHEMICALS INC Echinocystic acid 100MG
SDP
Catalog No. 502876529
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Echinocystic acid (EA), a natural triterpone enriched in various herbs, shows a range of pharmacological activities including anti-inflammatory and antioxidant effects. Purity 98.7%
Catalog No. 50-287-6529 Supplier TARGETMOL CHEMICALS INC Supplier No. T2866100MG
Add to Cart
Add to Cart
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Echinocystic acid (EA), a natural triterpone enriched in various herbs, shows a range of pharmacological activities including anti-inflammatory and antioxidant effects. Purity 98.7%