Learn More
Medchemexpress LLC Echinocystic acid 28-O-β-D-glucoside | 99633-30-4 | 1 MG
SDP
Catalog No. 5000417545
Encompass_Preferred
Echinocystic acid 28-O-β-D-glucoside is a metabolite of Echinocystic acid, formed through microbial oxidation and glucosidation. It functions as an inhibitor of the tissue factor pathway.
  • Acts as a tissue factor pathway inhibitor
  • Inhibits tissue factor procoagulant activities in human THP-1 cells
  • IC50 of 10.61 nM for tissue factor inhibition
Catalog No. 50-004-17545 Supplier Medchemexpress LLC Supplier No. HYN81021MG
Add to Cart
Add to Cart
Echinocystic acid 28-O-β-D-glucoside is a metabolite of Echinocystic acid, formed through microbial oxidation and glucosidation. It functions as an inhibitor of the tissue factor pathway.
  • Acts as a tissue factor pathway inhibitor
  • Inhibits tissue factor procoagulant activities in human THP-1 cells
  • IC50 of 10.61 nM for tissue factor inhibition