Learn More
Medchemexpress LLC Echinocystic acid 28-O-β-D-glucoside | 99633-30-4 | 5 MG
SDP
Catalog No. 5000417611
Encompass_Preferred
Echinocystic acid 28-O-β-D-glucoside is a metabolite of Echinocystic acid, formed through microbial oxidation and glucosidation. It acts as a potent tissue factor pathway inhibitor with an IC50 of 10.61 nM, effectively inhibiting tissue factor (TF) procoagulant activities in human THP-1 cells when stimulated by lipopolysaccharide.
  • Metabolite of Echinocystic acid
  • Inhibits tissue factor pathway with an IC50 of 10.61 nM
  • Reduces TF procoagulant activities in THP-1 cells
Catalog No. 50-004-17611 Supplier Medchemexpress LLC Supplier No. HYN81025MG
Add to Cart
Add to Cart
Echinocystic acid 28-O-β-D-glucoside is a metabolite of Echinocystic acid, formed through microbial oxidation and glucosidation. It acts as a potent tissue factor pathway inhibitor with an IC50 of 10.61 nM, effectively inhibiting tissue factor (TF) procoagulant activities in human THP-1 cells when stimulated by lipopolysaccharide.
  • Metabolite of Echinocystic acid
  • Inhibits tissue factor pathway with an IC50 of 10.61 nM
  • Reduces TF procoagulant activities in THP-1 cells