Learn More
3B Scientific™ Echinodermata, Bryozoa, Brachiopoda Slide Set
SDPSCI_ED
Catalog No. S95039L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S95039L 10/Pk.
1 options
Catalog No. S95039L Supplier 3B Scientific™ Supplier No. W13037
Add to Cart
Add to Cart
10 microscope slides
  • 1(d). Asterias, starfish, t.s. of ray
  • 2(e). Young starfish, horizontal sec.
  • 3(f). Asterias, starfish, bipinnaria larva, w.m.
  • 4(d). Echinus, young sea urchin, radial section
  • 5(e). Development of sea urchin, eggs in different stages
  • 6(d). Echinus, sea urchin, pluteus larva
  • 7(e). Holothuria, sea cucumber, t.s.
  • 8(c). Holothuria, w.m. of limy bodies
  • 9(d). Bryozoa, moss animals, colony, sec.
  • 10(f). Lingula, brachiopod, t.s.
Specifications
Type Echinodermata, Bryozoa and Brachiopoda
Quantity 10/Pk.