Learn More
Medchemexpress LLC Echinomycin (quinomycin A) | 512-64-1 | 98.6% | 1101.26 | C51H64N12O12S2 | 1 MG
SDP
Catalog No. 5000243721
Encompass_Preferred
Echinomycin is a cyclic peptide quinoxaline antibiotic used as a research reagent to probe hypoxia signaling and transcriptional regulation. It binds DNA and potently inhibits hypoxia-inducible factor-1 (HIF-1) DNA-binding activity, and has been reported to selectively affect cancer stem cell populations at low picomolar concentrations.
  • Potent inhibitor of HIF-1 DNA-binding activity.
  • High purity suitable for in vitro and cellular assays.
  • Stable as a solid when stored at low temperature under inert atmosphere.
  • Provides reproducible biological activity in hypoxia and transcription studies.
  • Available documentation includes datasheet and certificate of analysis.
Catalog No. 50-002-43721 Supplier Medchemexpress LLC Supplier No. HY1061011MG
Echinomycin is a cyclic peptide quinoxaline antibiotic used as a research reagent to probe hypoxia signaling and transcriptional regulation. It binds DNA and potently inhibits hypoxia-inducible factor-1 (HIF-1) DNA-binding activity, and has been reported to selectively affect cancer stem cell populations at low picomolar concentrations.
  • Potent inhibitor of HIF-1 DNA-binding activity.
  • High purity suitable for in vitro and cellular assays.
  • Stable as a solid when stored at low temperature under inert atmosphere.
  • Provides reproducible biological activity in hypoxia and transcription studies.
  • Available documentation includes datasheet and certificate of analysis.