Learn More
AdipoGen Echinosporin
SDP
Catalog No. 502562154
Encompass_Preferred
Chemical. CAS 79127-35-8. Formula C10H9NO5. MW 223.2. Isolated from Streptomyces sp. Antibiotic. Cell cycle inhibitor at the G2/M phase. Antitumor compound. Apoptosis inducer.
Catalog No. 50-256-2154 Supplier AdipoGen Supplier No. BVT0006M005
Add to Cart
Add to Cart
Chemical. CAS 79127-35-8. Formula C10H9NO5. MW 223.2. Isolated from Streptomyces sp. Antibiotic. Cell cycle inhibitor at the G2/M phase. Antitumor compound. Apoptosis inducer.