Learn More
AdipoGen Echinosporin
SDP
Supplier:  AdipoGen BVT0006M001
Encompass_Preferred
Chemical. CAS 79127-35-8. Formula C10H9NO5. MW 223.2. Isolated from Streptomyces sp. Antibiotic. Cell cycle inhibitor at the G2/M phase. Antitumor compound. Apoptosis inducer.
Catalog No. 50-256-2155