Learn More
Bachem Echistatin
SDP
Supplier:  Bachem 4030571.0100
Encompass_Preferred
Echistatin , H-Glu-Cys-Glu-Ser-Gly-Pro-Cys-Cys-Arg-Asn-Cys-Lys-Phe-Leu-Lys-Glu-Gly-Thr-Ile-Cys-Lys-Arg-Ala-Arg-Gly-Asp-Asp-Met-Asp-Asp-Tyr-Cys-Asn-Gly-Lys-Thr-Cys-Asp-Cys-Pro-Arg-Asn-Pro-His-Lys-Gly-Pro-Ala-Thr-OH, 154303-05-6, C217H341N71O74S9, Mr 5417.12, 0.1mg. Echistatin is a 49-amino acid polypeptide from the venom of the saw-scaled viper, Echis carinatus. The peptide containing an RGD motif binds to the glycoprotein IIb/IIIa receptor on the platelet and is a potent competitive inhibitor of ADPstimulated platelet aggregation mediated by fibrinogen and other factors. Furthermore, echistatin effectively inhibits osteoclastic bone resorption in vitro and in vivo.
Catalog No. 50-260-207