Learn More
Medchemexpress LLC Echitamine chloride | 6878-36-0 | 1 MG
SDP
Catalog No. 5000418331
Encompass_Preferred
Echitamine chloride, also known as Ditaine chloride, is a prominent monoterpene indole alkaloid found in Alstonia scholaris. It is known for its potent anti-tumour properties, operating by inducing DNA fragmentation and promoting cell apoptosis. Additionally, it acts as an inhibitor of pancreatic lipase.
  • Potent anti-tumor activity
  • Induces DNA fragmentation
  • Promotes cell apoptosis
  • Inhibits pancreatic lipase with an IC50 of 10.92 μM
Catalog No. 50-004-18331 Supplier Medchemexpress LLC Supplier No. HYN3797A1MG
Echitamine chloride, also known as Ditaine chloride, is a prominent monoterpene indole alkaloid found in Alstonia scholaris. It is known for its potent anti-tumour properties, operating by inducing DNA fragmentation and promoting cell apoptosis. Additionally, it acts as an inhibitor of pancreatic lipase.
  • Potent anti-tumor activity
  • Induces DNA fragmentation
  • Promotes cell apoptosis
  • Inhibits pancreatic lipase with an IC50 of 10.92 μM