Learn More
Beckman Coulter Echo qualified 1536-well low dead volume source microplate (COC), clear, barcoded - for Echo acoustic liquid handlers
Catalog No. NC1731974
Encompass
Echo-qualified 1536-well low dead volume source microplate constructed from cyclic olefin copolymer (COC) for acoustic liquid handling. Designed and lot tested to meet Echo transfer specifications, the plate provides optical clarity and reduced dead volume to support precise, low-volume transfers in fluorescence and luminescence assays.
  • Reduces dead volume and increases working range.
  • Provides optical clarity for luminescence and fluorescence assays.
  • Barcoded for automation and sample tracking.
  • Designed and lot tested for acoustic liquid transfer compatibility.
  • Compatible with heat seals and MicroClime environmental lids.
  • Packaged in anti-static bags and deionized to minimize contamination.
Catalog No. NC1731974 Supplier Beckman Coulter Supplier No. 0017913
Echo-qualified 1536-well low dead volume source microplate constructed from cyclic olefin copolymer (COC) for acoustic liquid handling. Designed and lot tested to meet Echo transfer specifications, the plate provides optical clarity and reduced dead volume to support precise, low-volume transfers in fluorescence and luminescence assays.
  • Reduces dead volume and increases working range.
  • Provides optical clarity for luminescence and fluorescence assays.
  • Barcoded for automation and sample tracking.
  • Designed and lot tested for acoustic liquid transfer compatibility.
  • Compatible with heat seals and MicroClime environmental lids.
  • Packaged in anti-static bags and deionized to minimize contamination.