Learn More
Beckman Coulter Echo Qualified 384-well Cyclic Olefin Copolymer (COC) Source Microplate, Low Dead Volume, Clear, Sterile, Surface-treated with Carboxyl Coating
Catalog No. NC1731976
Encompass
Echo Qualified 384-Well Low Dead Volume (LDV) Source Microplates are designed to minimize dead volume when using the Echo Liquid Handler. This microplate is constructed of Cyclic Olefin Copolymer (COC) for optimal acoustic transfer performance and has been specially optimized for high precision aqueous liquid transfers when using the Echo 525 Liquid Handler. Each lot of our coated source microplates is tested and certified to meet stringent transfer performance specifications on the Echo acoustic liquid handling platform. The 384LDV Echo Qualified Carboxyl Coated Source Microplate is distinguishable from the standard Echo Qualified 384-well Source Microplates by a yellow dot on the long side of the lower P-24 corner.
Catalog No. NC1731976 Supplier Beckman Coulter Supplier No. 00113070
Echo Qualified 384-Well Low Dead Volume (LDV) Source Microplates are designed to minimize dead volume when using the Echo Liquid Handler. This microplate is constructed of Cyclic Olefin Copolymer (COC) for optimal acoustic transfer performance and has been specially optimized for high precision aqueous liquid transfers when using the Echo 525 Liquid Handler. Each lot of our coated source microplates is tested and certified to meet stringent transfer performance specifications on the Echo acoustic liquid handling platform. The 384LDV Echo Qualified Carboxyl Coated Source Microplate is distinguishable from the standard Echo Qualified 384-well Source Microplates by a yellow dot on the long side of the lower P-24 corner.