Learn More
Beckman Coulter Qualified 384-Well Polypropylene Microplate, Clear, Non-sterile, 100/cs
Supplier:  Beckman Coulter 00114555
Encompass
Echo Qualified 384-Well Polypropylene (PP) Source Microplates ensure accurate and precise transfer with the Echo Liquid Handler. This microplate is highly resistant to solvents including DMSO and buffers making it ideal for compound and sample storage.
Catalog No. NC1731982