Learn More
Torrey Pines Scientific™ EchoTherm™ Cover
Catalog No. 1168033
Change view
Click to view available options
For Use With (Application):
Aluminum sample block
Catalog No. For Use With (Application)
11-680-33 Aluminum sample block
1 options
Catalog No. 11-680-33 Supplier Torrey Pines Scientific™ Supplier No. 7200009
Add to Cart
Add to Cart
For Use With: EchoTherm™ Benchtop Chilling/Heating Incubator
Clear covers constructed of vacuformed polyethylene. Choose from two designs: deep cover for use with sample blocks and shallow cover for use with 96-well assay plates without sample blocks. Both with handle for easier use.
Specifications
Type Cover
For Use With (Application) Aluminum sample block